accessibility menu, dialog, popup

UserName

FFFCl

CAS RN 21742-00-7

IUPAC Name: 1-(chloromethyl)-2-(trifluoromethyl)benzene
Synonyms: 1-(chloromethyl)-2-(trifluoromethyl)benzene
Molecular Weight (g/mol): 194.58
Molecular Formula: C8H6ClF3
InChi Key: BBXDMCQDLOCXRA-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=CC=C1CCl
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results