CAS RN 217487-19-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Menadione Sodium Bisulfite Hydrate, TRC
CAS: 217487-19-9 Molecular Formula: C11 H9 O5 S . H2 O . Na Molecular Weight (g/mol): 294.256 IUPAC Name: sodium;2-methyl-1,4-dioxo-3H-naphthalene-2-sulfonate;hydrate SMILES: O.[Na+].CC1(CC(=O)c2ccccc2C1=O)S(=O)(=O)[O-]