missing translation for 'onlineSavingsMsg'
Learn More

Menadione Sodium Bisulfite Hydrate, TRC

Product Code. 30484013
Change view
Click to view available options
Quantity:
5 g
10 g
50 g
Unit Size:
10g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30484013 50 g 50g
30481577 5 g 5g
30478086 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30484013 Supplier Toronto Research Chemicals Supplier No. TRCM21866350G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 217487-19-9
Molecular Formula C11 H9 O5 S . H2 O . Na
Molecular Weight (g/mol) 294.256
IUPAC Name sodium;2-methyl-1,4-dioxo-3H-naphthalene-2-sulfonate;hydrate
SMILES O.[Na+].CC1(CC(=O)c2ccccc2C1=O)S(=O)(=O)[O-]

Specifications

Quantity 50 g
InChI Formula InChI=1S/C11H10O5S.Na.H2O/c1-11(17(14,15)16)6-9(12)7-4-2-3-5-8(7)10(11)13;;/h2-5H,6H2,1H3,(H,14,15,16);;1H2/q;+1;/p-1
Formula Weight 294.017
Chemical Name or Material Menadione Sodium Bisulfite Hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.