CAS RN 21770-81-0
IUPAC Name:
carbamimidoyl(methyl)azanium chloride
Synonyms:
carbamimidoyl(methyl)azanium chloride
Molecular Weight (g/mol):
109.56
Molecular Formula:
C2H8ClN3
InChi Key:
VJQCNCOGZPSOQZ-UHFFFAOYSA-N
SMILES:
[Cl-].C[NH2+]C(N)=N
1
–
1
of
1
results
1
N-Methylguanidine Hydrochloride, TRC
CAS: 21770-81-0 Molecular Formula: C2H8ClN3 Molecular Weight (g/mol): 109.56 Synonym: N-Methylguanidine monohydrochloride,Methylguanidinium Chloride IUPAC Name: 1-methylguanidine;hydrochloride SMILES: Cl.CNC(=N)N