missing translation for 'onlineSavingsMsg'
Learn More

N-Methylguanidine Hydrochloride, TRC

Product Code. 30475742
Change view
Click to view available options
Quantity:
25 g
250 g
Unit Size:
250g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30475742 250 g 250g
30470969 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30475742 Supplier Toronto Research Chemicals Supplier No. TRCM311776250G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21770-81-0
Molecular Formula C2H8ClN3
Molecular Weight (g/mol) 109.56
Synonym N-Methylguanidine monohydrochloride, Methylguanidinium Chloride
IUPAC Name 1-methylguanidine;hydrochloride
SMILES Cl.CNC(=N)N

Specifications

Quantity 250 g
InChI Formula InChI=1S/C2H7N3.ClH/c1-5-2(3)4;/h1H3,(H4,3,4,5);1H
Formula Weight 109.0407
Chemical Name or Material N-Methylguanidine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.