accessibility menu, dialog, popup

UserName

CAS RN 2181-04-6

KH3C(R)O(R)(S)(S)H3C(S)(R)HOOO

CAS RN 2181-04-6

IUPAC Name: potassium 3-[(1R,3aS,3bR,9aR,9bS,11aS)-1-hydroxy-9a,11a-dimethyl-7-oxo-1H,2H,3H,3aH,3bH,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-yl]propanoate
Synonyms: potassium canrenoate
Molecular Weight (g/mol): 396.57
Molecular Formula: C22H29KO4
InChi Key: JTZQCHFUGHIPDF-RYVBEKKQSA-M
SMILES: [K+].C[C@]12CC[C@H]3[C@@H](C=CC4=CC(=O)CC[C@]34C)[C@@H]1CC[C@@]2(O)CCC([O-])=O
Molecular Weight (g/mol) 
  • (2)
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)