missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Canrenoic Acid Potassium Salt, TRC
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | Canrenoic Acid Potassium Salt |
| CAS | 2181-04-6 |
| Molecular Formula | C22 H29 K O4 |
| Quantity | 5 g |
| Synonym | (17α)-17-Hydroxy-3-oxopregna-4,6-diene-21-carboxylic Acid Potassium Salt, Aldadiene Kalium, Aldadiene Potassium, CB 8109, Canrenoate Potassium, Kanrenol, MF 465 a, Potassium 17-Hydroxy-3-oxo-17α-pregna-4,6-diene-21-carboxylate, Potassium 3-(17β-Hydroxy-3-oxoandrosta-4,6-dien-17-yl)propionate, Potassium 3-(3-oxo-17β-hydroxy-4,6-androstadien-17α-yl)propionate, Potassium Aldadiene, Potassium Canrenoate, Potassium β-(3-Oxo-17β-hydroxyandrosta-4,6-dien-17α-yl) Propionate, SC 14266, SC 14266/371, Soldactona, Soldactone, Soludactone, Venactone |
| InChI Formula | InChI=1S/C22H30O4.K/c1-20-9-5-15(23)13-14(20)3-4-16-17(20)6-10-21(2)18(16)7-11-22(21,26)12-8-19(24)25;/h3-4,13,16-18,26H,5-12H2,1-2H3,(H,24,25);/q;+1/p-1/t16-,17+,18+,20+,21+,22-;/m1./s1 |
| SMILES | [K+].C[C@]12CCC(=O)C=C1C=C[C@@H]3[C@@H]2CC[C@@]4(C)[C@H]3CC[C@@]4(O)CCC(=O)[O-] |
| IUPAC Name | potassium;3-[(8R,9S,10R,13S,14S,17R)-17-hydroxy-10,13-dimethyl-3-oxo-2,8,9,11,12,14,15,16-octahydro-1H-cyclopenta[a]phenanthren-17-yl]propanoate |
| Molecular Weight (g/mol) | 396.56 |
| Formula Weight | 396.1703 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction