accessibility menu, dialog, popup

UserName

POO

CAS RN 218282-19-0

IUPAC Name: 3-(triphenyl-λ⁵-phosphanylidene)oxan-2-one
Synonyms: 3-(triphenyl-λ⁵-phosphanylidene)oxan-2-one
Molecular Weight (g/mol): 360.39
Molecular Formula: C23H21O2P
InChi Key: PZGUKMFSICDTKX-UHFFFAOYSA-N
SMILES: O=C1OCCCC1=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results