accessibility menu, dialog, popup

UserName

H3CCH3H3COONNH2N

CAS RN 219503-81-8

IUPAC Name: tert-butyl 6-amino-1H-indazole-1-carboxylate
Synonyms: tert-butyl 6-aminoindazole-1-carboxylate
Molecular Weight (g/mol): 233.27
Molecular Formula: C12H15N3O2
InChi Key: ZUBZKAXFZUIBQY-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1N=CC2=CC=C(N)C=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results