missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-Boc-6-amino-1H-indazole, 97%

Product Code. 15454677
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15454677 250 mg 250mg
15444677 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15454677 Supplier Alfa Aesar™ Supplier No. H32010.MD
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Applications
Used in the synthesis of chiral molecules.

Solubility
Slightlysoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 219503-81-8
Molecular Formula C12H15N3O2
Molecular Weight (g/mol) 233.27
MDL Number MFCD04114657
InChI Key ZUBZKAXFZUIBQY-UHFFFAOYSA-N
Synonym 1-boc-6-amino-1h-indazole, tert-butyl 6-amino-1h-indazole-1-carboxylate, 1-boc-6-amino-indazole, 1-boc-6-aminoindazole, 6-amino-1h-indazole-1-carboxylic acid, 1,1 dimethyl ethyl ester, 1h-indazole-1-carboxylic acid, 6-amino-, 1,1-dimethylethyl ester, acmc-1cff2, ksc494o9t, 1-boc-1h-indazol-6-amine, 6-amino-1h-indazole, n1-boc protected
PubChem CID 12018607
IUPAC Name tert-butyl 6-aminoindazole-1-carboxylate
SMILES CC(C)(C)OC(=O)N1N=CC2=CC=C(N)C=C12

Specifications

Melting Point 171°C to 172°C
Quantity 250 mg
Solubility Information Slightlysoluble in water.
Formula Weight 233.27
Percent Purity 97%
Chemical Name or Material 1-Boc-6-amino-1H-indazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.