accessibility menu, dialog, popup

UserName

CAS RN 2196-13-6

IUPAC Name: pyridine-4-carbothioamide
Synonyms: 4-pyridinecarbothioamide
Molecular Weight (g/mol): 138.19
Molecular Formula: C6H6N2S
InChi Key: KPIIGXWUNXGGCP-UHFFFAOYSA-N
SMILES: NC(=S)C1=CC=NC=C1
Molecular Weight (g/mol) 
  • (3)
  • (5)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (5)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (3)
  • (2)