missing translation for 'onlineSavingsMsg'
Learn More

Thioisonicotinamide, 98%, Thermo Scientific™

Product Code. 11490818
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11490818 25 g 25g
11463930 1 g 1g
11480818 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11490818 Supplier Thermo Scientific Alfa Aesar Supplier No. B25265.14
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2196-13-6
Molecular Formula C6H6N2S
Molecular Weight (g/mol) 138.188
MDL Number MFCD00006437
InChI Key KPIIGXWUNXGGCP-UHFFFAOYSA-N
Synonym thioisonicotinamide, 4-pyridinecarbothioamide, isothionicotinamide, 4-pyridylthiocarboxamide, 4-pyridylthioamide, thio-4-pyridinecarboxamide, isonicotinic acid thioamide, pyridine-4-thioamide, 4-pyridinecarboxamide, thio, isonicotinamide, thio
PubChem CID 2723788
IUPAC Name pyridine-4-carbothioamide
SMILES C1=CN=CC=C1C(=S)N

Specifications

Melting Point 198°C to 202°C (decomposition)
Quantity 25 g
Beilstein 2174
Formula Weight 138.19
Percent Purity 98%
Chemical Name or Material Thioisonicotinamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.