accessibility menu, dialog, popup

UserName

H3COBrOO

CAS RN 2199-83-9

IUPAC Name: 6-bromo-3-butanoyl-2H-chromen-2-one
Synonyms: 6-bromo-3-butanoylchromen-2-one
Molecular Weight (g/mol): 295.13
Molecular Formula: C13H11BrO3
InChi Key: RDBKKAPCFCJFND-UHFFFAOYSA-N
SMILES: CCCC(=O)C1=CC2=CC(Br)=CC=C2OC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results