accessibility menu, dialog, popup

UserName

CAS RN 2216-12-8

ONOO

CAS RN 2216-12-8

IUPAC Name: 1-nitro-2-phenoxybenzene
Synonyms: benzene, 1-nitro-2-phenoxy-
Molecular Weight (g/mol): 215.21
Molecular Formula: C12H9NO3
InChi Key: VNHGETRQQSYUGZ-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=CC=C1OC1=CC=CC=C1
Boiling Point 
  • (3)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results