accessibility menu, dialog, popup

UserName

CH3NNH

CAS RN 22261-94-5

IUPAC Name: 1-methyl-N-phenylpiperidin-4-amine
Synonyms: 1-methyl-N-phenylpiperidin-4-amine
Molecular Weight (g/mol): 190.29
Molecular Formula: C12H18N2
InChi Key: GXJSWUKLJQHQFZ-UHFFFAOYSA-N
SMILES: CN1CCC(CC1)NC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results