missing translation for 'onlineSavingsMsg'
Learn More

4-Anilino-1-methylpiperidine, 98%, Thermo Scientific™

Product Code. 11362319
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11362319 25 g 25g
11352319 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11362319 Supplier Thermo Scientific Alfa Aesar Supplier No. H27276.14
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22261-94-5
Molecular Formula C12H18N2
Molecular Weight (g/mol) 190.29
MDL Number MFCD06408133
InChI Key GXJSWUKLJQHQFZ-UHFFFAOYSA-N
PubChem CID 89642
IUPAC Name 1-methyl-N-phenylpiperidin-4-amine
SMILES CN1CCC(CC1)NC2=CC=CC=C2

Specifications

Melting Point 82°C to 86°C
Quantity 25 g
Synonym 4-anilino-1-methylpiperidine, n-1-methylpiperidin-4-yl aniline, 4-piperidinamine,1-methyl-n-phenyl, 1-methyl 4-piperidyl phenylamine, acmc-20akdg, 4-piperidinamine, 1-methyl-n-phenyl, 4-anilino-1-methylpiperidin, 4-anilino-1-methyl-piperidine, n-1-methyl-4-piperidinyl aniline, 1-methyl-n-phenyl-4-piperidinamine
Formula Weight 190.28
Percent Purity 98%
Chemical Name or Material 4-Anilino-1-methylpiperidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.