accessibility menu, dialog, popup

UserName

CH3SOOCl

CAS RN 22381-53-9

IUPAC Name: 1-(2-chloroethanesulfonyl)-4-methylbenzene
Synonyms: 1-(2-chloroethanesulfonyl)-4-methylbenzene
Molecular Weight (g/mol): 218.7
Molecular Formula: C9H11ClO2S
InChi Key: KUAGAJLNIOIGRN-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)S(=O)(=O)CCCl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results