CAS RN 2239-67-0
CAS RN 2239-67-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
H-Gly-Pro-Hyp-OH, TRC
CAS: 2239-67-0 Molecular Formula: C12H19N3O5 Molecular Weight (g/mol): 285.3 Synonym: 1: PN: WO2007017671 PAGE: 49 Claimed Protein,26: PN: 3WO2014175556 TABLE: 5 Claimed Sequence,70: PN: WO03006603 SEQID: 76 Claimed Protein,9: PN: WO2017020025 SEQID: 9 Claimed Sequence,Glycylprolylhydroxyproline,Tripeptide 29; IUPAC Name: (2S,4R)-1-[(2S)-1-(2-aminoacetyl)pyrrolidine-2-carbonyl]-4-hydroxypyrrolidine-2-carboxylic acid SMILES: NCC(=O)N1CCC[C@H]1C(=O)N2C[C@H](O)C[C@H]2C(=O)O