missing translation for 'onlineSavingsMsg'
Learn More

H-Gly-Pro-Hyp-OH, TRC

Product Code. 30240868
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30240868 25 mg 25mg
30240501 5 mg 5mg
30240617 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30240868 Supplier Toronto Research Chemicals Supplier No. TRCG65014325MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2239-67-0
Molecular Formula C12H19N3O5
Molecular Weight (g/mol) 285.3
Synonym 1: PN: WO2007017671 PAGE: 49 Claimed Protein, 26: PN: 3WO2014175556 TABLE: 5 Claimed Sequence, 70: PN: WO03006603 SEQID: 76 Claimed Protein, 9: PN: WO2017020025 SEQID: 9 Claimed Sequence, Glycylprolylhydroxyproline, Tripeptide 29;
IUPAC Name (2S,4R)-1-[(2S)-1-(2-aminoacetyl)pyrrolidine-2-carbonyl]-4-hydroxypyrrolidine-2-carboxylic acid
SMILES NCC(=O)N1CCC[C@H]1C(=O)N2C[C@H](O)C[C@H]2C(=O)O

Specifications

InChI Formula InChI=1S/C12H19N3O5/c13-5-10(17)14-3-1-2-8(14)11(18)15-6-7(16)4-9(15)12(19)20/h7-9,16H,1-6,13H2,(H,19,20)/t7-,8+,9+/m1/s1
Quantity 25 mg
Formula Weight 285.1325
Percent Purity >95%
Shipping Condition Room Temperature
Chemical Name or Material H-Gly-Pro-Hyp-OH
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.