accessibility menu, dialog, popup

UserName

NH2BrNOHN

CAS RN 2240-25-7

IUPAC Name: 6-amino-5-bromo-1,2-dihydropyrimidin-2-one
Synonyms: 4-amino-5-bromo-3H-pyrimidin-2-one
Molecular Weight (g/mol): 190
Molecular Formula: C4H4BrN3O
InChi Key: QFVKLKDEXOWFSL-UHFFFAOYSA-N
SMILES: NC1=C(Br)C=NC(=O)N1
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results