missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-5-bromo-2-hydroxypyrimidine, 98%

Product Code. 15458538
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15458538 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15458538 Supplier Thermo Scientific Alfa Aesar Supplier No. H61952.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 2240-25-7 | C4H4BrN3O | 190.00 g/mol

4-Amino-5-bromo-2-hydroxypyrimidine can be used in the synthesis of cross-link products under anaerobic and aerobic conditions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Amino-5-bromo-2-hydroxypyrimidine can be used in the synthesis of cross-link products under anaerobic and aerobic conditions.

Solubility
Soluble in formic acid (50 mg/ml).

Notes
Store away from st
TRUSTED_SUSTAINABILITY

Specifications

CAS 2240-25-7
Melting Point 240°C to 243°C
Color White to Yellow
Molecular Formula C4H4BrN3O
MDL Number MFCD00056025
Quantity 1 g
Beilstein 127286
Synonym 5-bromocytosine, 4-amino-5-bromopyrimidin-2 1h-one, 6-amino-5-bromopyrimidin-2 1h-one, 4-amino-5-bromo-2-pyrimidinol, 4-amino-5-bromopyrimidin-2-ol, 2 1h-pyrimidinone, 4-amino-5-bromo, 4-amino-5-bromo-2-hydroxypyrimidine, 2 1h-pyrimidinone,6-amino-5-bromo, 6-amino-5-bromo-1,2-dihydropyrimidin-2-one, 4-amino-5-bromo-1,2-dihydropyrimidin-2-one
Solubility Information Soluble in formic acid (50mg/ml).
InChI Key QFVKLKDEXOWFSL-UHFFFAOYSA-N
SMILES NC1=C(Br)C=NC(=O)N1
IUPAC Name 6-amino-5-bromo-1H-pyrimidin-2-one
Molecular Weight (g/mol) 190.00
PubChem CID 75233
Formula Weight 190
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 4-Amino-5-bromo-2-hydroxypyrimidine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.