accessibility menu, dialog, popup

UserName

CAS RN 224185-19-7

CH3NOOBrF

CAS RN 224185-19-7

IUPAC Name: 1-bromo-2-fluoro-4-methyl-5-nitrobenzene
Synonyms: 1-bromo-2-fluoro-4-methyl-5-nitrobenzene
Molecular Weight (g/mol): 234.02
Molecular Formula: C7H5BrFNO2
InChi Key: MTTNTJRJOFVWSY-UHFFFAOYSA-N
SMILES: CC1=CC(F)=C(Br)C=C1[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results