accessibility menu, dialog, popup

UserName

CAS RN 2244-16-8

IUPAC Name: (5S)-2-methyl-5-(prop-1-en-2-yl)cyclohex-2-en-1-one
Synonyms: carvone, (+)- (S)-(+)-carvone d-p-mentha-6,8(9)-dien-2-one d-(+)-carvone (+)-carvone D-(+)-carvone D(+)-carvone d-carvone d-carvone (natural) (+)-(S)-carvone
Molecular Weight (g/mol): 150.22
Molecular Formula: C10H14O
InChi Key: ULDHMXUKGWMISQ-VIFPVBQESA-N
SMILES: CC(=C)[C@H]1CC=C(C)C(=O)C1
Molecular Weight (g/mol) 
  • (7)
  • (2)
Quantity 
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
Boiling Point 
  • (2)
  • (4)
Percent Purity 
  • (4)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (7)
  • (2)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (2)
  • (4)
  • (4)
  • (2)
  • (4)