missing translation for 'onlineSavingsMsg'
Learn More

D(+)-Carvone, 96%, natural

Product Code. 10664622 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10664622 25 g Glass bottle 25g
10674622 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10664622 Supplier Thermo Scientific Acros Supplier No. 150680250
 Restricted Item
Estimated Shipment: 20-10-2026
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

D(+)-Carvone, CAS # 2244-16-8, is a monoterpene ketone, a natural product found mainly in Achillea ageratum and Curcuma pierreana. It can be used as a flavoring agent. | CAS: 2244-16-8 | C10H14O | 150.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • D(+)-Carvone belongs to terpene family and is considered a monoterpene ketone naturally found mainly in Achillea ageratum and Curcuma pierreana

Application

  • D(+)-Carvone has shown a suppressant effect against high-fat diet induced weight gain in mice
  • It has been used as a flavoring agent
  • It has been used in organic synthesis as starting material for the asymmetric total synthesis of natural products, including the synthesis of the terpenoid quassin
  • In vitro studies demonstrated that this compound can inhibit cellulase-induced capsidiol production in Nicotiana tabacum by interfering with protein isoprenylation
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2244-16-8
Molecular Formula C10H14O
Molecular Weight (g/mol) 150.22
MDL Number MFCD00062997
InChI Key ULDHMXUKGWMISQ-VIFPVBQESA-N
Synonym d-carvone, +-carvone, s-+-carvone, s-carvone, s-2-methyl-5-prop-1-en-2-yl cyclohex-2-enone, +-s-carvone, carvone, +, d +-carvone, carvol, s-+-p-mentha-6,8-dien-2-one
PubChem CID 16724
ChEBI CHEBI:15399
IUPAC Name (5S)-2-methyl-5-prop-1-en-2-ylcyclohex-2-en-1-one
SMILES CC1=CCC(CC1=O)C(=C)C

Specifications

Color Colorless to Yellow
Density 0.9560g/mL
Boiling Point 230.0°C
Flash Point 97°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Refractive Index 1.4950 to 1.5010
Quantity 25 g
Beilstein 07, 153
Merck Index 14, 1874
Specific Gravity 0.956
Specific Rotation + 57.00
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol, ether, chloroform,, propylene glycol and mineral oils, insoluble in glycerol
Formula Weight 150.22
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material D(+)-Carvone, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.