accessibility menu, dialog, popup

UserName

CAS RN 2251-65-2

IUPAC Name: 3-(trifluoromethyl)benzoyl chloride
Synonyms: 3-(trifluoromethyl)benzoyl chloride
Molecular Weight (g/mol): 208.56
Molecular Formula: C8H4ClF3O
InChi Key: RUJYJCANMOTJMO-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=CC(=C1)C(Cl)=O
Boiling Point 
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)