accessibility menu, dialog, popup

UserName

NNClCl

CAS RN 22536-67-0

IUPAC Name: 2,5-dichloropyrimidine
Synonyms: 2,5-dichloropyrimidine
Molecular Weight (g/mol): 148.97
Molecular Formula: C4H2Cl2N2
InChi Key: CEJAHXLRNZJPQH-UHFFFAOYSA-N
SMILES: ClC1=CN=C(Cl)N=C1
Molecular Weight (g/mol) 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)