missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dichloropyrimidine, 95%, Thermo Scientific™

Product Code. 15264318 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15264318 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15264318 Supplier Thermo Scientific Acros Supplier No. 454040010
 Restricted Item
Estimated Shipment: 31-07-2026
Add to basket
This item is not returnable. View return policy

Specifications

CAS 22536-67-0
Melting Point 52.0°C to 55.0°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass Bottle
Molecular Formula C4H2Cl2N2
Quantity 1 g
Synonym 2,5-dichloro-pyrimidine, pyrimidine, 2,5-dichloro, 2,5-dichloro-1,3-diazine, pubchem13600, acmc-209fxc, ksc493a8n, 2,5-dichloropyrimidine, pyrimidine, 2,5-dichloro-7ci,8ci,9ci
InChI Key CEJAHXLRNZJPQH-UHFFFAOYSA-N
SMILES C1=C(C=NC(=N1)Cl)Cl
IUPAC Name 2,5-dichloropyrimidine
Molecular Weight (g/mol) 148.98
PubChem CID 590641
Formula Weight 148.98
Percent Purity 95%
Chemical Name or Material 2,5-Dichloropyrimidine
Show More Show Less
compliance-icons
Product Identifier
  • 2,5-Dichloropyrimidine, 95%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.