accessibility menu, dialog, popup

UserName

LiBHCH3H3CH3C

CAS RN 22560-16-3

IUPAC Name: lithium(1+) triethylboranuide
Synonyms: lithium triethylborohydride
Molecular Weight (g/mol): 105.94
Molecular Formula: C6H16BLi
InChi Key: WCJAYABJWDIZAJ-UHFFFAOYSA-N
SMILES: [Li+].CC[BH-](CC)CC
Quantity 
  • (4)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (5)
  • (4)
  • (2)
  • (1)
  • (1)
  • (5)