accessibility menu, dialog, popup

UserName

CAS RN 2265-91-0

FIF

CAS RN 2265-91-0

IUPAC Name: 1,3-difluoro-5-iodobenzene
Synonyms: 1,3-difluoro-5-iodobenzene
Molecular Weight (g/mol): 239.99
Molecular Formula: C6H3F2I
InChi Key: QQCFOQNFPIAENW-UHFFFAOYSA-N
SMILES: FC1=CC(I)=CC(F)=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results