accessibility menu, dialog, popup

UserName

FFFBr

CAS RN 226717-83-5

IUPAC Name: 1-(bromomethyl)-2,3,5-trifluorobenzene
Synonyms: 1-(bromomethyl)-2,3,5-trifluorobenzene
Molecular Weight (g/mol): 225.01
Molecular Formula: C7H4BrF3
InChi Key: QPMAEHBAXJSICW-UHFFFAOYSA-N
SMILES: FC1=CC(F)=C(F)C(CBr)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results