accessibility menu, dialog, popup

UserName

CAS RN 2269-22-9

AlH3CH3COH3CCH3OH3CCH3O

CAS RN 2269-22-9

IUPAC Name: aluminium(3+) tris(butan-2-olate)
Synonyms: aluminum butoxide
Molecular Weight (g/mol): 246.33
Molecular Formula: C12H27AlO3
InChi Key: WOZZOSDBXABUFO-UHFFFAOYNA-N
SMILES: [Al+3].CCC(C)[O-].CCC(C)[O-].CCC(C)[O-]
Molecular Weight (g/mol) 
  • (3)
  • (9)
Percent Purity 
  • (6)
  • (6)
Quantity 
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (4)
Boiling Point 
  • (6)
  • (3)
Physical Form 
  • (6)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (2)
  • (9)
  • (3)
  • (9)
  • (6)
  • (6)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (4)
  • (6)
  • (3)
  • (6)
  • (3)