missing translation for 'onlineSavingsMsg'
Learn More

Aluminum tri-sec-butoxide, 97%

Product Code. 10405365 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Unit Size:
100g
2.5kg
500g
This item is not returnable. View return policy

Product Code. 10405365

Brand: Thermo Scientific Acros 191211000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2269-22-9 | C12H27AlO3 | 246.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2269-22-9
Molecular Formula C12H27AlO3
Molecular Weight (g/mol) 246.33
MDL Number MFCD00009327
InChI Key LWGPRERTOLVRLK-UHFFFAOYSA-N
Synonym aluminum sec-butoxide, aluminum tri-sec-butoxide, 2-butanol, aluminum salt, aluminum sec-butylate, aluminum butan-2-olate, aluminum tri-s-butoxide, 2-butanol, aluminum salt 3:1, aluminum tri-sec-butanolate, al o-sec-bu 3, aluminiumtri-sec-butoxide
PubChem CID 50931103
IUPAC Name aluminum;butan-2-ol
SMILES CCC(C)O.CCC(C)O.CCC(C)O.[Al]

Specifications

Color Yellow
Density 0.9600g/mL
Boiling Point 200.0°C to 206.0°C (30.0 mmHg)
Flash Point 27°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (ex Al)
Packaging Glass bottle
Linear Formula [C2H5CH(CH3)O]3Al
Refractive Index 1.4380 to 1.4400
Quantity 100 g
Beilstein 01,IV,1567
Specific Gravity 0.96
Solubility Information Solubility in water: decomposes
Viscosity 1800 mPa.s (20°C)
Formula Weight 246.33
Percent Purity 97%
Physical Form Oily Liquid
Chemical Name or Material Aluminum tri-sec-butoxide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Aluminium tri-sec-butoxide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.