accessibility menu, dialog, popup

UserName

CH3ONHSCH3OO

CAS RN 22821-80-3

IUPAC Name: N-(4-methanesulfonylphenyl)acetamide
Synonyms: N-(4-methanesulfonylphenyl)acetamide
Molecular Weight (g/mol): 213.25
Molecular Formula: C9H11NO3S
InChi Key: SJYUABJSWXGSAO-UHFFFAOYSA-N
SMILES: CC(=O)NC1=CC=C(C=C1)S(C)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results