accessibility menu, dialog, popup

UserName

CAS RN 23000-14-8

IUPAC Name: dimethyl-1,3-oxazole-4-carboxylic acid
Synonyms: dimethyl-1,3-oxazole-4-carboxylic acid
Molecular Weight (g/mol): 141.13
Molecular Formula: C6H7NO3
InChi Key: LHGRUGVXZLHYKE-UHFFFAOYSA-N
SMILES: CC1=NC(C(O)=O)=C(C)O1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results