accessibility menu, dialog, popup

UserName

H3CH3CCH3OONHOHOCl

CAS RN 232275-73-9

IUPAC Name: 4-{[(tert-butoxy)carbonyl]amino}-2-chlorobenzoic acid
Synonyms: 4-[(tert-butoxycarbonyl)amino]-2-chlorobenzoic acid
Molecular Weight (g/mol): 271.7
Molecular Formula: C12H14ClNO4
InChi Key: HDYBNYDKCZJPPX-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NC1=CC=C(C(O)=O)C(Cl)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results