accessibility menu, dialog, popup

UserName

CAS RN 232595-59-4

IUPAC Name: 3-{[(tert-butoxy)carbonyl]amino}-5-hydroxybenzoic acid
Synonyms: 3-[(tert-butoxycarbonyl)amino]-5-hydroxybenzoic acid
Molecular Weight (g/mol): 253.25
Molecular Formula: C12H15NO5
InChi Key: KSTCXQVSGVVIAO-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NC1=CC(O)=CC(=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results