accessibility menu, dialog, popup

UserName

CAS RN 2344-83-4

CH3SiCH3H3CCl

CAS RN 2344-83-4

IUPAC Name: (3-chloropropyl)trimethylsilane
Synonyms: (3-chloropropyl)trimethylsilane
Molecular Weight (g/mol): 150.72
Molecular Formula: C6H15ClSi
InChi Key: QXDDDCNYAAJLBT-UHFFFAOYSA-N
SMILES: C[Si](C)(C)CCCCl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results