CAS RN 23491-45-4
CAS RN 23491-45-4
IUPAC Name:
trihydrogen 4-{5-[6-(4-methylpiperazin-1-yl)-1H-1,3-benzodiazol-2-yl]-2,3-dihydro-1H-1,3-benzodiazol-2-ylidene}cyclohexa-2,5-dien-1-one trichloride
Synonyms:
trihydrogen 4-{5-[5-(4-methylpiperazin-1-yl)-3H-1,3-benzodiazol-2-yl]-1,3-dihydro-1,3-benzodiazol-2-ylidene}cyclohexa-2,5-dien-1-one trichloride
Molecular Weight (g/mol):
533.88
Molecular Formula:
C25H27Cl3N6O
InChi Key:
OKQAASMPIOSOEX-UHFFFAOYSA-N
SMILES:
[H+].[H+].[H+].[Cl-].[Cl-].[Cl-].CN1CCN(CC1)C1=CC=C2N=C(NC2=C1)C1=CC=C2NC(NC2=C1)=C1C=CC(=O)C=C1
1
–
1
of
1
results
1
MedChemExpress Hoechst 33258 trihydrochloride
MedChemExpress Hoechst 33258 trihydrochloride is a fluorescent dyes, which can be used as a cell dye for DNA.