accessibility menu, dialog, popup

UserName

CH3BrHO

CAS RN 2362-12-1

IUPAC Name: 4-bromo-2-methylphenol
Synonyms: 2-methyl-4-bromophenol
Molecular Weight (g/mol): 187.04
Molecular Formula: C7H7BrO
InChi Key: IWJGMJHAIUBWKT-UHFFFAOYSA-N
SMILES: CC1=CC(Br)=CC=C1O
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results