accessibility menu, dialog, popup

UserName

H3CBr

CAS RN 23703-22-2

IUPAC Name: 1-bromo-4-hexylbenzene
Synonyms: 1-bromo-4-hexylbenzene
Molecular Weight (g/mol): 241.17
Molecular Formula: C12H17Br
InChi Key: MXHOLIARBWJKCR-UHFFFAOYSA-N
SMILES: CCCCCCC1=CC=C(Br)C=C1
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)