accessibility menu, dialog, popup

UserName

CAS RN 2373-76-4

CH3HOO(E)

CAS RN 2373-76-4

IUPAC Name: (2E)-3-(2-methylphenyl)prop-2-enoic acid
Synonyms: (2E)-3-(2-methylphenyl)prop-2-enoic acid
Molecular Weight (g/mol): 162.19
Molecular Formula: C10H10O2
InChi Key: RSWBWHPZXKLUEX-VOTSOKGWSA-N
SMILES: CC1=CC=CC=C1\C=C\C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results