accessibility menu, dialog, popup

UserName

CAS RN 2386-54-1

NaH3CSOOO

CAS RN 2386-54-1

IUPAC Name: sodium butane-1-sulfonate
Synonyms: sodium 1-butanesulfonate
Molecular Weight (g/mol): 160.16
Molecular Formula: C4H9NaO3S
InChi Key: XQCHMGAOAWZUPI-UHFFFAOYSA-M
SMILES: [Na+].CCCCS([O-])(=O)=O
Quantity 
  • (3)
  • (5)
  • (2)
Percent Purity 
  • (2)
  • (3)
  • (4)
Physical Form 
  • (6)
  • (1)
Grade 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (4)
  • (3)
  • (3)
  • (5)
  • (2)
  • (2)
  • (3)
  • (4)
  • (6)
  • (1)
  • (2)
  • (2)