missing translation for 'onlineSavingsMsg'
Learn More

1-Butane Sulfonic Acid, Sodium Salt, for HPLC, Certified, Fisher Chemical

Product Code. 10191510 Shop All Fisher Chemical Products
Change view
Click to view available options
:
25g, Plastic powder jar
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10191510 25g, Plastic powder jar
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10191510 Supplier Fisher Chemical Supplier No. B/4791/46
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Melting Point 300°C
CAS 2386-54-1
Color White
Physical Form Powder/Solid
Quantity 25 g
Molecular Formula C4H9NaO3S
MDL Number 7540
InChI Key XQCHMGAOAWZUPI-UHFFFAOYSA-M
SMILES [Na+].CCCCS([O-])(=O)=O
IUPAC Name sodium butane-1-sulfonate
Molecular Weight (g/mol) 160.16
Formula Weight 160.17g/mol
Packaging Plastic powder jar
Show More Show Less
compliance-icons
Product Identifier
  • 1-Butanesulfonic acid, sodium salt
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.