accessibility menu, dialog, popup

UserName

CAS RN 23886-64-8

BrSOOClBr

CAS RN 23886-64-8

IUPAC Name: 2,5-dibromobenzene-1-sulfonyl chloride
Synonyms: 2,5-dibromobenzenesulfonyl chloride
Molecular Weight (g/mol): 334.41
Molecular Formula: C6H3Br2ClO2S
InChi Key: ZLMPLIWURYRGEB-UHFFFAOYSA-N
SMILES: ClS(=O)(=O)C1=C(Br)C=CC(Br)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results