missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dibromobenzenesulfonyl chloride, 99%

Product Code. 11399905
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11399905 5 g 5g
11389905 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11399905 Supplier Thermo Scientific Alfa Aesar Supplier No. L19888.06
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 23886-64-8 | C6H3Br2ClO2S | 334.406 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23886-64-8
Molecular Formula C6H3Br2ClO2S
Molecular Weight (g/mol) 334.406
MDL Number MFCD00060685
InChI Key ZLMPLIWURYRGEB-UHFFFAOYSA-N
Synonym 2,5-dibromobenzene-1-sulfonyl chloride, 2,5-dibromobenzenesulphonyl chloride, benzenesulfonyl chloride, 2,5-dibromo, 2,5-dibromophenyl chlorosulfone, 2,5-dibromobenzene sulphonyl chloride, pubchem5112, acmc-1cbvd, ksc201m8p, 2,5-dibromophenylsulfonyl chloride, 2,5-dibromobenzenesulphonylchloride
PubChem CID 2737515
IUPAC Name 2,5-dibromobenzenesulfonyl chloride
SMILES C1=CC(=C(C=C1Br)S(=O)(=O)Cl)Br

Specifications

Melting Point 69°C to 72°C
Quantity 5 g
UN Number UN3261
Beilstein 2694054
Sensitivity Moisture sensitive
Formula Weight 334.41
Percent Purity 99%
Chemical Name or Material 2,5-Dibromobenzenesulfonyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.