accessibility menu, dialog, popup

UserName

CAS RN 2390-59-2

ClCH3NCH3CNCH3H3CNCH3H3C

CAS RN 2390-59-2

IUPAC Name: tris[4-(diethylamino)phenyl]methylium chloride
Synonyms: tris[4-(diethylamino)phenyl]methylium chloride
Molecular Weight (g/mol): 492.15
Molecular Formula: C31H42ClN3
InChi Key: JVICFMRAVNKDOE-UHFFFAOYSA-M
SMILES: [Cl-].CCN(CC)C1=CC=C(C=C1)[C+](C1=CC=C(C=C1)N(CC)CC)C1=CC=C(C=C1)N(CC)CC
Quantity 
  • (1)
  • (3)
  • (1)
Color 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (1)
  • (2)