missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Violet, MP Biomedicals

Product Code. 11425441 Shop All MP Biomedicals Products
Click to view available options
Quantity:
25 g
Unit Size:
25g

Product Code. 11425441

Brand: MP Biomedicals 0215801625

Please to purchase this item. Need a web account? Register with us today!

Specifications

CAS 2390-59-2
Assay Percent Range ∽80%
Molecular Formula C31H42ClN3
MDL Number MFCD00011765
Synonym Basic Violet 4, Ethyl purple 6B
InChI Key JVICFMRAVNKDOE-UHFFFAOYSA-M
SMILES [Cl-].CCN(CC)C1=CC=C(C=C1)[C+](C1=CC=C(C=C1)N(CC)CC)C1=CC=C(C=C1)N(CC)CC
IUPAC Name tris[4-(diethylamino)phenyl]methylium chloride
Molecular Weight (g/mol) 492.15
PubChem CID 16955
Formula Weight 492.1
Quantity 25 g
Chemical Name or Material Ethyl Violet
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.