accessibility menu, dialog, popup

UserName

CAS RN 239087-07-1

FFFFBr

CAS RN 239087-07-1

IUPAC Name: 1-(bromomethyl)-2-fluoro-4-(trifluoromethyl)benzene
Synonyms: 1-(bromomethyl)-2-fluoro-4-(trifluoromethyl)benzene
Molecular Weight (g/mol): 257.03
Molecular Formula: C8H5BrF4
InChi Key: MQUCRCQNRDHRPL-UHFFFAOYSA-N
SMILES: FC1=C(CBr)C=CC(=C1)C(F)(F)F
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results