accessibility menu, dialog, popup

UserName

FBrFFF

CAS RN 239087-08-2

IUPAC Name: 2-(bromomethyl)-1-fluoro-3-(trifluoromethyl)benzene
Synonyms: 2-(bromomethyl)-1-fluoro-3-(trifluoromethyl)benzene
Molecular Weight (g/mol): 257.03
Molecular Formula: C8H5BrF4
InChi Key: RINUERVPFANASB-UHFFFAOYSA-N
SMILES: FC1=CC=CC(=C1CBr)C(F)(F)F
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)