CAS RN 2396-68-1
IUPAC Name:
4-tert-butylbenzene-1-thiol
Synonyms:
benzenethiol, p-tert-butyl-
Molecular Weight (g/mol):
166.28
Molecular Formula:
C10H14S
InChi Key:
GNXBFFHXJDZGEK-UHFFFAOYSA-N
SMILES:
CC(C)(C)C1=CC=C(S)C=C1
1
–
1
of
1
results
1
4-tert-Butylthiophenol, 97%
CAS: 2396-68-1 Molecular Formula: C10H14S Molecular Weight (g/mol): 166.282 MDL Number: MFCD00022067 InChI Key: GNXBFFHXJDZGEK-UHFFFAOYSA-N Synonym: 4-tert-butylthiophenol,p-tert-butylthiophenol,benzenethiol, p-tert-butyl,p-tert-butylbenzenethiol,p-t-butylthiophenol,p-tert-butylphenyl mercaptan,benzenethiol, 4-1,1-dimethylethyl,benzenethiol, p-tert-butylthio,4-tert-butyl thiophenol,4-t-butylthiophenol PubChem CID: 75454 IUPAC Name: 4-tert-butylbenzenethiol SMILES: CC(C)(C)C1=CC=C(C=C1)S